accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4316-98-7

NH2NNClH2N

CAS RN 4316-98-7

IUPAC Name: 6-chloropyrimidine-4,5-diamine
Synonyms: 6-chloropyrimidine-4,5-diamine
Molecular Weight (g/mol): 144.56
Molecular Formula: C4H5ClN4
InChi Key: VNSFICAUILKARD-UHFFFAOYSA-N
SMILES: NC1=NC=NC(Cl)=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results