accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOOOO

CAS RN 439571-48-9

IUPAC Name: bis[(2H-1,3-benzodioxol-5-yl)methyl][(1-butyl-2,4-diphenyl-1H-imidazol-5-yl)methyl]amine
Synonyms: bis(2H-1,3-benzodioxol-5-ylmethyl)[(3-butyl-2,5-diphenylimidazol-4-yl)methyl]amine
Molecular Weight (g/mol): 573.69
Molecular Formula: C36H35N3O4
InChi Key: ITACCRHKSPSKKL-UHFFFAOYSA-N
SMILES: CCCCN1C(CN(CC2=CC3=C(OCO3)C=C2)CC2=CC3=C(OCO3)C=C2)=C(N=C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)