Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NDT 9513727
GSA_VAGREENER_CHOICESDP

Catalog No. 333350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-335-0 50 mg
33-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-335-0 Supplier R&D Systems™ Supplier No. 3333/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective human C5a receptor negative allosteric modulator (NAM)

Potent C5a receptor (C5aR) negative allosteric modulator (NAM) (IC50 = 11.6 nM); selective for human C5aR (IC50 values for rat and mouse C5aR are > 10 μM). Exhibits inverse agonist pharmacokinetics. Blocks degranulation response in C5a-stimulated U937 cells (IC50 = 7.1 nM). Exhibits selectivity for primate and gerbil receptor over other species. Orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 439571-48-9
Molecular Formula C36H35N3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 573.68
Purity >98%
Molecular Weight (g/mol) 573.693
Chemical Name or Material NDT 9513727
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.