accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4403-69-4

IUPAC Name: 2-(aminomethyl)aniline
Synonyms: 2-(aminomethyl)aniline
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: GVOYKJPMUUJXBS-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)