accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClFNNOHNN

CAS RN 443144-27-2

IUPAC Name: hydrogen 7-{4-[2-(4-fluorophenyl)ethyl]piperazine-1-carbonyl}-1H-indole-3-carbonitrile chloride
Synonyms: hydrogen pruvanserin chloride
Molecular Weight (g/mol): 412.89
Molecular Formula: C22H22ClFN4O
InChi Key: UYGNZRVLPYZLTF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].FC1=CC=C(CCN2CCN(CC2)C(=O)C2=C3NC=C(C#N)C3=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results