accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNSNOH

CAS RN 443292-81-7

IUPAC Name: N-(4-ethoxyphenyl)-4-(hydroxydiphenylmethyl)piperidine-1-carbothioamide
Synonyms: N-(4-ethoxyphenyl)-4-(hydroxydiphenylmethyl)piperidine-1-carbothioamide
Molecular Weight (g/mol): 446.61
Molecular Formula: C27H30N2O2S
InChi Key: PMEQBGAGFZDWQX-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(=S)N2CCC(CC2)C(O)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)