CAS RN 443776-49-6
IUPAC Name:
{4-[(carbamimidoylsulfanyl)methyl]phenyl}boronic acid hydrobromide
Synonyms:
4-[(carbamimidoylsulfanyl)methyl]phenylboronic acid hydrobromide
Molecular Weight (g/mol):
290.97
Molecular Formula:
C8H12BBrN2O2S
InChi Key:
PAFZAMOVHIRQOD-UHFFFAOYSA-N
SMILES:
Br.NC(=N)SCC1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
Tocris Bioscience™ BC 11 hydrobromide
CAS: 443776-49-6 Molecular Formula: C8H12BBrN2O2S