accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4447-60-3

IUPAC Name: 2-[bis(propan-2-yloxy)methoxy]propane
Synonyms: 2-(diisopropoxymethoxy)propane
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: FPIVAWNGRDHRSQ-UHFFFAOYSA-N
SMILES: CC(C)OC(OC(C)C)OC(C)C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)