Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropyl Orthoformate 97.0+%, TCI America™
SDP

Catalog No. O021525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O021525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O021525ML Supplier TCI America Supplier No. O021525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4447-60-3
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.283
MDL Number MFCD00060628
InChI Key FPIVAWNGRDHRSQ-UHFFFAOYSA-N
Synonym triisopropyl orthoformate, triisopropoxymethane, triisopropylorthoformate, propane, 2,2',2-methylidynetris oxy tris, 2-diisopropoxymethoxy propane, 2-di propan-2-yloxy methoxy propane, 2-bis propan-2-yloxy methoxy propane, orthoformic acid, triisopropyl ester, isopropyl orthoformate
PubChem CID 78191
IUPAC Name 2-[di(propan-2-yloxy)methoxy]propane
SMILES CC(C)OC(OC(C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 65°C
Quantity 25 mL
UN Number 3272
Formula Weight 190.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropyl Orthoformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.