accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3ONHNNOHNN

CAS RN 444723-13-1

IUPAC Name: 4-{[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino}-N,N-diethylbenzamide
Synonyms: 4-{[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino}-N,N-diethylbenzamide
Molecular Weight (g/mol): 422.53
Molecular Formula: C23H30N6O2
InChi Key: XHEQSRJCJTWWAH-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(NC2=NC(OCC3CCCCC3)=C3NC=NC3=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)