Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NU 6140
GREENER_CHOICESDP

Catalog No. 330110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 33-011-0 Supplier R&D Systems™ Supplier No. 3301/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cdk2 inhibitor; also potent inhibitor of aurora kinase A and B

Cyclin-dependent kinase 2 (cdk2) inhibitor (IC50 values are 0.42 - 1.1 μM). Displays some selectivity for cdk2 (IC50 value = 0.94 μM for cdk4 and >2.3 μM for other cdks). Induces cell-cycle arrest at the G2-M phase. Potentiates the apoptotic effect of taxol (Cat. No. 1097) in HeLa cells; down-regulates the antiapoptotic protein survivin. Also potently inhibits aurora kinases (IC50 values are 0.035 and 0.067 μM for aurora kinase B and A, respectively).

Chemical Identifiers

CAS 444723-13-1
Molecular Formula C23H30N6O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 422.52
Purity >99%
Molecular Weight (g/mol) 422.533
Chemical Name or Material NU 6140
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.