accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOO

CAS RN 4455-09-8

IUPAC Name: 2-phenylethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-phenylethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 276.35
Molecular Formula: C15H16O3S
InChi Key: CVPPUZPZPFOFPK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results