accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOPOOHHONH2

CAS RN 449173-19-7

IUPAC Name: {2-amino-2-[(3-octylphenyl)carbamoyl]ethoxy}phosphonic acid
Synonyms: 2-amino-2-[(3-octylphenyl)carbamoyl]ethoxyphosphonic acid
Molecular Weight (g/mol): 372.4
Molecular Formula: C17H29N2O5P
InChi Key: MRUSUGVVWGNKFE-UHFFFAOYNA-N
SMILES: CCCCCCCCC1=CC(NC(=O)C(N)COP(O)(O)=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results