accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHHHClClClClNH2NH2NH2H2N

CAS RN 4506-66-5

IUPAC Name: tetrahydrogen benzene-1,2,4,5-tetramine tetrachloride
Synonyms: tetrahydrogen benzene-1,2,4,5-tetramine tetrachloride
Molecular Weight (g/mol): 284.01
Molecular Formula: C6H14Cl4N4
InChi Key: BZDGCIJWPWHAOF-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NC1=CC(N)=C(N)C=C1N
Quantity 
  • (1)
  • (1)
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results