accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHONNHNN

CAS RN 452342-67-5

IUPAC Name: N-(oxan-4-yl)-4-{4-[5-(pyridin-2-yl)-1H-pyrazol-4-yl]pyridin-2-yl}benzamide
Synonyms: N-(oxan-4-yl)-4-{4-[3-(pyridin-2-yl)-2H-pyrazol-4-yl]pyridin-2-yl}benzamide
Molecular Weight (g/mol): 425.49
Molecular Formula: C25H23N5O2
InChi Key: SAGZIBJAQGBRQA-UHFFFAOYSA-N
SMILES: O=C(NC1CCOCC1)C1=CC=C(C=C1)C1=NC=CC(=C1)C1=C(NN=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results