CAS RN 459147-39-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ SW 033291
High affinity 15-PGDH inhibitor; promotes hematopoiesis and hepatocyte proliferation