accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOHNCl

CAS RN 459168-41-3

IUPAC Name: 5-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-indole
Synonyms: 5-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-indole
Molecular Weight (g/mol): 277.75
Molecular Formula: C14H16ClN3O
InChi Key: HUQJRYMLJBBEDO-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C(=O)C1=CC2=CC(Cl)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)