accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4602-70-4

H3CONCH3HO

CAS RN 4602-70-4

IUPAC Name: 6-methoxy-1-methyl-3,4-dihydroisoquinolin-7-ol
Synonyms: 6-methoxy-1-methyl-3,4-dihydroisoquinolin-7-ol
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: GNFURDSMULTPOS-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1O)C(C)=NCC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results