accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NONHOOOH3CH3CCH3NHOH3C

CAS RN 461054-93-3

IUPAC Name: tert-butyl 3-[14-methoxy-2-(2-methylpropyl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraen-5-yl]propanoate
Synonyms: tert-butyl 3-[14-methoxy-2-(2-methylpropyl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraen-5-yl]propanoate
Molecular Weight (g/mol): 469.58
Molecular Formula: C26H35N3O5
InChi Key: NXNRAECHCJZNRF-UHFFFAOYNA-N
SMILES: COC1=CC=C2C(NC3=C2CC2N(C3CC(C)C)C(=O)C(CCC(=O)OC(C)(C)C)NC2=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results