accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4659-45-4

IUPAC Name: 2,6-dichlorobenzoyl chloride
Synonyms: 2,6-dichlorobenzoyl chloride
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: JBLIDPPHFGWTKU-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C=CC=C1Cl
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)