CAS RN 4722-94-5
IUPAC Name:
4-chloropyridine-2,6-dicarboxylic acid
Synonyms:
4-chloropyridine-2,6-dicarboxylic acid
Molecular Weight (g/mol):
201.56
Molecular Formula:
C7H4ClNO4
InChi Key:
IYUMNONNHYADBU-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(Cl)=CC(=N1)C(O)=O
1
–
1
of
1
results
1
4-Chloro-2,6-pyridinedicarboxylic Acid 98.0+%, TCI America™
CAS: 4722-94-5 Molecular Formula: C7H4ClNO4 Molecular Weight (g/mol): 201.562 MDL Number: MFCD04117894 InChI Key: IYUMNONNHYADBU-UHFFFAOYSA-N PubChem CID: 304010 IUPAC Name: 4-chloropyridine-2,6-dicarboxylic acid SMILES: C1=C(C=C(N=C1C(=O)O)C(=O)O)Cl