accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOCl(E)

CAS RN 472981-92-3

IUPAC Name: (2E)-3-(4-chlorophenyl)-N-(3-methoxyphenyl)prop-2-enamide
Synonyms: (2E)-3-(4-chlorophenyl)-N-(3-methoxyphenyl)prop-2-enamide
Molecular Weight (g/mol): 287.74
Molecular Formula: C16H14ClNO2
InChi Key: RYAMDQKWNKKFHD-JXMROGBWSA-N
SMILES: COC1=CC=CC(NC(=O)\C=C\C2=CC=C(Cl)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)