Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SB 366791
GREENER_CHOICESDP

Catalog No. 161550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-155-0 50 mg
16-151-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-155-0 Supplier R&D Systems™ Supplier No. 1615/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective, competitive TRPV1 antagonist

Potent, selective and competitive vanilloid TRPV1 receptor antagonist (pA2 = 7.71 at hVR1); antagonizes hTRPV1 receptors activated by agonists, noxious heat, but not protons. Displays selectivity over a wide range of receptors and systems including CB1 and CB2 receptors, voltage-gated Ca2+ channels and the hyperpolarization-activated current (Ih).

Chemical Identifiers

CAS 472981-92-3
Molecular Formula C16H14ClNO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 287.75
Purity >99%
Molecular Weight (g/mol) 287.743
Chemical Name or Material SB 366791
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.