accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONONNH3CNNOFFFF

CAS RN 473722-68-8

IUPAC Name: N-{1-[3-(4-ethoxyphenyl)-4-oxo-3H,4H-pyrido[2,3-d]pyrimidin-2-yl]ethyl}-2-[4-fluoro-3-(trifluoromethyl)phenyl]-N-[(pyridin-3-yl)methyl]acetamide
Synonyms: N-{1-[3-(4-ethoxyphenyl)-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl}-2-[4-fluoro-3-(trifluoromethyl)phenyl]-N-(pyridin-3-ylmethyl)acetamide
Molecular Weight (g/mol): 605.59
Molecular Formula: C32H27F4N5O3
InChi Key: XMRGQUDUVGRCBS-UHFFFAOYNA-N
SMILES: CCOC1=CC=C(C=C1)N1C(=O)C2=CC=CN=C2N=C1C(C)N(CC1=CN=CC=C1)C(=O)CC1=CC(=C(F)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)