accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4744-08-5

IUPAC Name: 1,1-diethoxypropane
Synonyms: propane, 1,1-diethoxy-
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: MBNMGGKBGCIEGF-UHFFFAOYSA-N
SMILES: CCOC(CC)OCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)