Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionaldehyde Diethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. P0781500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0781500ML 500 mL
P078125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0781500ML Supplier TCI America Supplier No. P0781500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4744-08-5
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.20
MDL Number MFCD00009245
InChI Key MBNMGGKBGCIEGF-UHFFFAOYSA-N
Synonym propionaldehyde diethyl acetal, propane, 1,1-diethoxy, propanaldiethylacetal, propionaldehyde, diethyl acetal, diethyl propional, propanal diethyl acetal, propionaldehyde diethylacetal, propane,1-diethoxy, propioaldehyde diethylacetal, propioaldehyde diethyl acetal
PubChem CID 20858
IUPAC Name 1,1-diethoxypropane
SMILES CCC(OCC)OCC

Specifications

Color Colorless
Boiling Point 124°C
Quantity 500 mL
UN Number 3271
Formula Weight 132.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propionaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.