CAS RN 475193-31-8
IUPAC Name:
{[({5-[6-amino-2-(methylsulfanyl)-9H-purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy)(hydroxy)phosphoryl]oxy}phosphonic acid trisodium
Synonyms:
({5-[6-amino-2-(methylsulfanyl)purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy(hydroxy)phosphoryl)oxyphosphonic acid trisodium
Molecular Weight (g/mol):
542.26
Molecular Formula:
C11H17N5Na3O10P2S
InChi Key:
BIJRFSXCZAGVHQ-UHFFFAOYNA-N
SMILES:
[Na].[Na].[Na].CSC1=NC(N)=C2N=CN(C3OC(COP(O)(=O)OP(O)(O)=O)C(O)C3O)C2=N1
1
–
1
of
1
results
1
Tocris Bioscience™ 2-Methylthioadenosine diphosphate trisodium salt
CAS: 475193-31-8 Molecular Formula: C11H17N5Na3O10P2S