accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480424-78-0

NMgBr

CAS RN 480424-78-0

IUPAC Name: 1-{[4-(bromomagnesio)phenyl]methyl}pyrrolidine
Synonyms: 1-{[4-(bromomagnesio)phenyl]methyl}pyrrolidine
Molecular Weight (g/mol): 264.45
Molecular Formula: C11H14BrMgN
InChi Key: SMCUOBSDAGTZFC-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C(CN2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results