accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COCH3

CAS RN 4819-75-4

IUPAC Name: 1,1-diethoxy-2-methoxyethane
Synonyms: 1,1-diethoxy-2-methoxyethane
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: PCYADPMXIILFTP-UHFFFAOYSA-N
SMILES: CCOC(COC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results