accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSFONNNO

CAS RN 483313-22-0

IUPAC Name: 2-({1-[5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carbonyl]pyrrolidin-2-yl}methyl)-5-phenyl-1,3,4-oxadiazole
Synonyms: 2-({1-[5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carbonyl]pyrrolidin-2-yl}methyl)-5-phenyl-1,3,4-oxadiazole
Molecular Weight (g/mol): 448.52
Molecular Formula: C24H21FN4O2S
InChi Key: HYBZWVLPALMACV-UHFFFAOYNA-N
SMILES: CC1=NC(C(=O)N2CCCC2CC2=NN=C(O2)C2=CC=CC=C2)=C(S1)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)