accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHHO

CAS RN 487-49-0

IUPAC Name: 1-(2,4-dihydroxyphenyl)-2-(4-methoxyphenyl)ethan-1-one
Synonyms: 1-(2,4-dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 258.27
Molecular Formula: C15H14O4
InChi Key: XHBZOAYMBBUURD-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(=O)C2=CC=C(O)C=C2O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)