accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)OH(R)OH(S)OH(S)OHOH

CAS RN 488-44-8

IUPAC Name: (2R,3R,4S,5S)-hexane-1,2,3,4,5,6-hexol
Synonyms: (2R,3R,4S,5S)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol): 182.17
Molecular Formula: C6H14O6
InChi Key: FBPFZTCFMRRESA-FBXFSONDSA-N
SMILES: OC[C@H](O)[C@H](O)[C@H](O)[C@H](O)CO
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)