accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-84-6

IUPAC Name: (3R,4R)-2-(hydroxymethyl)oxolane-2,3,4-triol
Synonyms: ribulose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: LQXVFWRQNMEDEE-ZZKAVYKESA-N
SMILES: OCC1(O)OC[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)