accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHOCH3

CAS RN 489402-47-3

IUPAC Name: N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-2-carboxamide
Synonyms: N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-2-carboxamide
Molecular Weight (g/mol): 257.38
Molecular Formula: C17H23NO
InChi Key: SIQGKPGBLYKQBB-UHFFFAOYNA-N
SMILES: CC1=CC(C)=C(NC(=O)C2CC3CCC2C3)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results