CAS RN 492-02-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CC4
High affinity and subtype-selective α6β2 and α4β2 nAChR partial agonist