Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CC4
GREENER_CHOICESDP

Catalog No. 523650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-365-0 50 mg
52-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-365-0 Supplier Tocris Bioscience™ Supplier No. 5236/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and subtype-selective α6β2 and α4β2 nAChR partial agonist

CC4 is a high affinity and subtype selective α6β2 and α4β2 partial agonist (Ki values are 12 and 26nM for rat α6β2 and α4β2 receptors respectively). Has low affinity for α3β4 and α7 receptors (Ki values are 4.8 and 13 μM for human α3β4 and rat α7 receptors respectively). Stimulates dopamine release from striatal slices in vitro. Attenuates nicotine-induced self-administration and conditional place preference in rats.

Specifications

Chemical Name or Material (1R,5S)-1,2,3,4,5,6-Hexahydro-3-[2-[(1R,5S)-1,5,6,8-tetrahydro-8-oxo-1,5-methano-2H-pyrido[1,2-a][1,5]diazocin-3(4H)-yl]ethyl]-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one
CAS 492-02-4
Quantity 50 mg
Target Additional Nicotinic (a4b2) Receptor Compounds
Molecular Formula C24H30N4O2
Formula Weight 406.52
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.