accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONHNH3CHNOFFFHO(E/Z)(E/Z)

CAS RN 497259-23-1

IUPAC Name: N-[5-(hydroxymethyl)-1-methyl-3-oxo-2-(propan-2-yl)-1,2,3,4,5,6-hexahydro-1,4-benzodiazocin-8-yl]-5-[4-(trifluoromethyl)phenyl]penta-2,4-dienamide
Synonyms: N-[5-(hydroxymethyl)-2-isopropyl-1-methyl-3-oxo-2,4,5,6-tetrahydro-1,4-benzodiazocin-8-yl]-5-[4-(trifluoromethyl)phenyl]penta-2,4-dienamide
Molecular Weight (g/mol): 501.55
Molecular Formula: C27H30F3N3O3
InChi Key: WOLVEMPZUIFSII-UHFFFAOYNA-N
SMILES: CC(C)C1N(C)C2=C(CC(CO)NC1=O)C=C(NC(=O)C=CC=CC1=CC=C(C=C1)C(F)(F)F)C=C2
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results