Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TPPB
GREENER_CHOICESDP

Catalog No. 53431 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-431 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 53-431 Supplier R&D Systems™ Supplier No. 5343/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity PKC activator; also APP modulator

High affinity protein kinase C activator (Ki =11.9 nM). Induces differentiation of hESCs into Pdx-1 expressing pancreatic progenitor cells. Also enhances secretion of amyloid precursor protein. Cell permeable.

Chemical Identifiers

CAS 497259-23-1
Molecular Formula C27H30F3N3O3

Specifications

Quantity 1 mg
Solubility Soluble to 50mM in DMSO
Product Line Tocris
Formula Weight 501.54
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 501.55
Chemical Name or Material (2E,4E)-N-[(2S,5S)-1,2,3,4,5,6-Hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3-oxo-1,4-benz
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.