accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)O(S)(S)(R)OHOHO(S)CH3(S)HO(S)

CAS RN 50-23-7

IUPAC Name: (1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: hydrocortisone hydrocortisone alcohol hydrocortisonum hidrocortisona [inn-spanish] cortef cleiton hydrocortisone valerate idrocortisone hydrocortisone free alcohol 17α-hydroxycorticosterone
Molecular Weight (g/mol): 362.47
Molecular Formula: C21H30O5
InChi Key: JYGXADMDTFJGBT-VWUMJDOOSA-N
SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Molecular Weight (g/mol) 
  • (7)
  • (10)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (6)
  • (4)
Percent Purity 
  • (6)
  • (6)
Grade 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (1)
  • (5)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (6)
  • (7)
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
  • (6)
  • (4)
  • (6)
  • (6)
  • (1)
  • (6)