Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Resveratrol
GREENER_CHOICESDP

Catalog No. 1418100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-181-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-181-00 Supplier R&D Systems™ Supplier No. 1418/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cyclooxygenase inhibitor

A phytoestrogen with antitumor, antioxidant, antiplatelet, anti-inflammatory and antifungal effects. Inhibits cytochrome P450 1A1 (IC50 = 23 μM) and displays mixed agonist/antagonist actions at ERα and ERβ estrogen receptors. Converted into the anticancer agent piceatannol (Cat. No. 1554) by cytochrome P450 1B1. Activates autophagy. Also activates TRPA1 in prostate cancer-associated fibroblasts. Also SIRT1 activator.

Chemical Identifiers

CAS 501-36-0
Molecular Formula C14H12O3

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 228.25
Purity >98%
Molecular Weight (g/mol) 228.247
Chemical Name or Material Resveratrol
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.