accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHONHClOHCl

CAS RN 501684-93-1

IUPAC Name: 1-(5-chloro-2-hydroxyphenyl)-3-[2-chloro-5-(trifluoromethyl)phenyl]urea
Synonyms: 1-(5-chloro-2-hydroxyphenyl)-3-[2-chloro-5-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol): 365.13
Molecular Formula: C14H9Cl2F3N2O2
InChi Key: OUDXRNQPVSMGDW-UHFFFAOYSA-N
SMILES: OC1=C(NC(=O)NC2=C(Cl)C=CC(=C2)C(F)(F)F)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)