accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOONHOHOOH

CAS RN 501919-59-1

IUPAC Name: 2-hydroxy-4-{2-[(4-methylbenzenesulfonyl)oxy]acetamido}benzoic acid
Synonyms: 2-hydroxy-4-{2-[(4-methylbenzenesulfonyl)oxy]acetamido}benzoic acid
Molecular Weight (g/mol): 365.36
Molecular Formula: C16H15NO7S
InChi Key: HWNUSGNZBAISFM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(=O)NC1=CC(O)=C(C=C1)C(O)=O
Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results