Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NSC 74859
GREENER_CHOICESDP

Catalog No. 465550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-555-0 50 mg
46-551-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-555-0 Supplier R&D Systems™ Supplier No. 4655/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective STAT3 inhibitor

NSC 74859 selectively inhibits STAT3 DNA-binding activity in vitro (IC50 values are 86, 160, and >300 μM for STAT3-STAT3, STAT1-STAT3 and STAT1-STAT1 DNA-binding activity respectively). NSC 74859 blocks growth and induces apoptosis preferentially in tumor cells that contain persistently active STAT3 in vitro and arrests tumor growth in a murine xenograft model in vivo.

Chemical Identifiers

CAS 501919-59-1
Molecular Formula C16H15NO7S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 365.36
Purity >98%
Molecular Weight (g/mol) 365.356
Chemical Name or Material NSC 74859
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.