accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONNClCl

CAS RN 50264-69-2

IUPAC Name: 1-[(2,4-dichlorophenyl)methyl]-1H-indazole-3-carboxylic acid
Synonyms: DICA LND 氯尼达明 lonidaminum lonidamine lonidamina lonidamin doridamina diclondazolic acid
Molecular Weight (g/mol): 321.16
Molecular Formula: C15H10Cl2N2O2
InChi Key: WDRYRZXSPDWGEB-UHFFFAOYSA-N
SMILES: OC(=O)C1=NN(CC2=CC=C(Cl)C=C2Cl)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)