Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Lonidamine
GREENER_CHOICESDP

Catalog No. 164650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-465-0 50 mg
16-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-465-0 Supplier Tocris Bioscience™ Supplier No. 1646/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mitochondrial hexokinase inhibitor

Anticancer and antispermatogenic agent in vitro and in vivo. Inhibits cellular energy metabolism in some cells via inhibition of mitochondrial hexokinase. Also blocks CFTR Cl- channels in vitro.

Chemical Identifiers

CAS 50264-69-2
Molecular Formula C15H10Cl2N2O2

Specifications

Quantity 50 mg
Formula Weight 321.16
Purity >98%
Molecular Weight (g/mol) 321.157
Chemical Name or Material Lonidamine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.