accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNO(E/Z)

CAS RN 504433-23-2

IUPAC Name: 3-[(1-methyl-1H-indol-3-yl)methylidene]-1H,2H,3H-pyrrolo[3,2-b]pyridin-2-one
Synonyms: 3-[(1-methylindol-3-yl)methylidene]-1H-pyrrolo[3,2-b]pyridin-2-one
Molecular Weight (g/mol): 275.31
Molecular Formula: C17H13N3O
InChi Key: NXNQLECPAXXYTR-UHFFFAOYSA-N
SMILES: CN1C=C(C=C2C(=O)NC3=C2N=CC=C3)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results