accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CIAuI

CAS RN 50481-03-3

IUPAC Name: diiodogolduide; tetrabutylazanium
Synonyms: diiodogolduide; tetrabutylammonium ion
Molecular Weight (g/mol): 693.25
Molecular Formula: C16H36AuI2N
InChi Key: IPICULSEVGYZBD-UHFFFAOYSA-L
SMILES: I[Au-]I.CCCC[N+](CCCC)(CCCC)CCCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results