accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 506-48-9

IUPAC Name: octacosanoic acid
Synonyms: octacosanoic acid octaeicosanoic acid n-octaeicosanoic acid octacosansaeure montanic acid octacosancarbonsaeure
Molecular Weight (g/mol): 424.75
Molecular Formula: C28H56O2
InChi Key: UTOPWMOLSKOLTQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)