accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHOH2NOH2NF

CAS RN 507475-17-4

IUPAC Name: 2-(carbamoylamino)-5-(4-fluorophenyl)thiophene-3-carboxamide
Synonyms: 2-(carbamoylamino)-5-(4-fluorophenyl)thiophene-3-carboxamide
Molecular Weight (g/mol): 279.29
Molecular Formula: C12H10FN3O2S
InChi Key: SAYGKHKXGCPTLX-UHFFFAOYSA-N
SMILES: NC(=O)NC1=C(C=C(S1)C1=CC=C(F)C=C1)C(N)=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results