Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TPCA-1
GREENER_CHOICESDP

Catalog No. 255910 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-591-0 Supplier Tocris Bioscience™ Supplier No. 2559/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective inhibitor of IKKβ

Potent, selective inhibitor of IκB kinase (IKK) β (IC50 = 17.9 nM) that displays > 22-fold selectivity over IKKα and > 550-fold selectivity over other kinases and enzymes. Inhibits production of pro-inflammatory cytokines in vitro and in vivo and inhibits NF-κB nuclear localization. Reduces the severity and onset of collagen-induced arthritis; anti-inflammatory.

Chemical Identifiers

CAS 507475-17-4
Molecular Formula C12H10FN3O2S

Specifications

Quantity 10 mg
Formula Weight 279.29
Purity >97%
Molecular Weight (g/mol) 279.289
Chemical Name or Material TPCA-1
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.